Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-cis-Verbenol, 97%, sum of isomers, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18881-04-4
C10H16O
152.24
MFCD00065444
WONIGEXYPVIKFS-YIZRAAEISA-N
s-cis-verbenol,verbenol, s-cis,unii-xr9t57f48t,cis-verbenol,bicyclo 3.1.1 hept-3-en-2-ol, 4,6,6-trimethyl-, 1s,2s,5s,1s-cis-verbenol,ccris 5289,?-cis-verbenol
87839
(1S,2S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
CC1=CC(C2CC1C2(C)C)O
18881-04-4
C10H16O
152.24
MFCD00065444
WONIGEXYPVIKFS-YIZRAAEISA-N
s-cis-verbenol,verbenol, s-cis,unii-xr9t57f48t,cis-verbenol,bicyclo 3.1.1 hept-3-en-2-ol, 4,6,6-trimethyl-, 1s,2s,5s,1s-cis-verbenol,ccris 5289,?-cis-verbenol
87839
(1S,2S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-en-2-ol
CC1=CC(C2CC1C2(C)C)O
Melting Point 62°C to 68°C
Flash Point 87°C
Specific Rotation + 10
Refractive Index 1.49
Density 85g/mL
Color Bordeaux to Brown-Red
Quantity 25g
Packaging Glass bottle
Specific Rotation Condition + 10.00 (20.00°C c=2,C2H5OH)
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (sum of isomers) (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 152.24
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (S)-cis-Verbenol, Sum of isomers
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 242-645-2