Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ AT-100 Bulk Dispenser

Catalog Number p-89487
Click to view available options
Color:
Amber
Clear
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy

Specially designed to hold unpackaged PPE such as ambidextrous gloves, bouffant caps, beard covers, or ear plugs. Amber and gray dispensers help to protect latex gloves from ozone degradation.

  • Designed to dispense bulk PPE such as gloves, bouffant caps, or ear plugs
  • Unique access hole effectively dispenses a wide varitey of products
  • Amber and gray options help prevent ozone degradation of latex gloves
  • Can be mounted to the wall or free-standing
  • Single lid with full-length hinge for extra durability
  • Sloped top maintains HEPA flow
  • Flame treated edges to ensure a smooth finish
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 6 x 12 x 16 in.
Holds Gloves, Bouffant Caps, Ear Plugs
Material Acrylic
No. of Compartments 1
Product Style Top Loading
For Use With (Application) Dispensing and storing bulk PPE
Thickness (English) 0.25 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit