Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ X-Large Bouffant Dispenser

$254.50

Specifications

Color Clear
Dimensions (D x W x H) Interior 6 x 12 x 16 in.
For Use With (Application) Holds large volumes of bouffants, beard covers, etc
Includes Lid and oval front opening
Material PETG
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
19-162-567
View Documents
S-Curve™
BD616 PETG
PETG
Each for $254.50
Add to cart
 
19-162-409
View Documents
S-Curve™
BD616
Acrylic This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

Clear
Holds large volumes of bouffants, beard covers, etc
PETG
XL
6 x 12 x 16 in.
Lid and oval front opening
1
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit