Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Epichlorohydrin, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
67843-74-7
C3H5ClO
92.522
MFCD00077760
BRLQWZUYTZBJKN-GSVOUGTGSA-N
s-+-epichlorohydrin,s-epichlorohydrin,2s-2-chloromethyl oxirane,s-chloromethyl oxirane,s-3-chloropropylene oxide,s-2-chloromethyl oxirane,epichlorohydrin, +,unii-scr89b4r6o,ccris 6388
149428
CHEBI:37145
(2S)-2-(chloromethyl)oxirane
C1C(O1)CCl
67843-74-7
C3H5ClO
92.522
MFCD00077760
BRLQWZUYTZBJKN-GSVOUGTGSA-N
s-+-epichlorohydrin,s-epichlorohydrin,2s-2-chloromethyl oxirane,s-chloromethyl oxirane,s-3-chloropropylene oxide,s-2-chloromethyl oxirane,epichlorohydrin, +,unii-scr89b4r6o,ccris 6388
149428
CHEBI:37145
(2S)-2-(chloromethyl)oxirane
C1C(O1)CCl
Specific Rotation Condition + 34.00 (20.00°C 589nm,c=1,MeOH)
Color Colorless to Yellow
Quantity 1g
Density 1.1830g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4365 to 1.4385 (20°C, 589nm)
Specific Rotation +31° to +37° (20°C, 589nm) (c=1, MeOH)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 92.52
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material (S)-(+)-Epichlorohydrin
Show More Show Less

GHS H Statement:
May cause an allergic skin reaction.
Toxic if swallowed.
May cause cancer.
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger