Alfa Aesar™ S-Ethyl thioacetate, 98+%

Manufacturer: Alfa Aesar™ A1068022

Quantity 100g

 View more versions of this product

Catalog No. AAA1068022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 625-60-5
    Molecular Formula C4H8OS
    Formula Weight 104.18
    MDL Number MFCD00015178
    UN Number UN1993
    Beilstein 1737643
    Synonym Thioacetic acid S-ethyl ester
    Chemical Name or Material S-Ethyl thioacetate

    Specifications

    Formula Weight 104.18
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H8OS
    MDL Number MFCD00015178
    Beilstein 1737643
    Solubility Information Insoluble in water
    Boiling Point 116° to 117°C
    Quantity 100g
    Odor Stench
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 20°C (68°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 625-60-5
    UN Number UN1993
    Synonym Thioacetic acid S-ethyl ester
    Health Hazard 1 Danger
    Chemical Name or Material S-Ethyl thioacetate
    Assay ≥98%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit