Alfa Aesar™ S-Ethyl thiopropionate, 97%

Manufacturer: Alfa Aesar™ B2491806

Quantity 5g

 View more versions of this product

Catalog No. AAB2491806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2432-42-0
    Molecular Formula C5H10OS
    Formula Weight 118.2
    MDL Number MFCD00027016
    UN Number UN1993
    Beilstein 1740740
    Synonym Thiopropionic acid S-ethyl ester
    Chemical Name or Material S-Ethyl thiopropionate

    Specifications

    Formula Weight 118.2
    UN Number UN1993
    Health Hazard 3 GHS P Statement
    P210-P261
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Flash Point 27°C (80°F)
    Molecular Formula C5H10OS
    MDL Number MFCD00027016
    Beilstein 1740740
    Synonym Thiopropionic acid S-ethyl ester
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapour.
    Boiling Point 137° to 138°C
    Quantity 5g
    Odor Stench
    CAS 2432-42-0
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material S-Ethyl thiopropionate
    Assay 97%

    GHS H Statement
    H226
    Flammable liquid and vapour.

    GHS P Statement
    P210-P261
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit