Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ S-Ethyl thiopropionate, 97%

Click to view available options
Quantity:
5g
25g
100g
2432-42-0
C5H10OS
118.194
MFCD00027016
HNEVHBHRLCAKKQ-UHFFFAOYSA-N
s-ethyl thiopropionate,ethyl thiopropionate,propanethioic acid, s-ethyl ester,thiopropionic acid s-ethyl ester,1-ethylsulfanyl propan-1-one,ethanethiol propionate,acmc-1ceeh,propanethioicacid,s-ethylester,thiopropionic acid, s-ethyl ester,propanethioic acid,s-ethyl ester
75513
S-ethyl propanethioate
CCC(=O)SCC
2432-42-0
C5H10OS
118.194
MFCD00027016
HNEVHBHRLCAKKQ-UHFFFAOYSA-N
s-ethyl thiopropionate,ethyl thiopropionate,propanethioic acid, s-ethyl ester,thiopropionic acid s-ethyl ester,1-ethylsulfanyl propan-1-one,ethanethiol propionate,acmc-1ceeh,propanethioicacid,s-ethylester,thiopropionic acid, s-ethyl ester,propanethioic acid,s-ethyl ester
75513
S-ethyl propanethioate
CCC(=O)SCC
Boiling Point 137°C to 138°C
Quantity 25g
Flash Point 27°C (80°F)
Odor Stench
UN Number UN1993
Beilstein 1740740
Formula Weight 118.2
Percent Purity 97%
Chemical Name or Material S-Ethyl thiopropionate

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P261
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.