Alfa Aesar™ S-Furfuryl thioacetate, 99%

Manufacturer: Alfa Aesar™ B2182114

Quantity 25g

 View more versions of this product

Catalog No. AAB2182114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13678-68-7
    Molecular Formula C7H8O2S
    Formula Weight 156.2
    MDL Number MFCD00010083
    UN Number UN3334
    Synonym Thioacetic acid S-furfuryl ester
    Chemical Name or Material S-Furfuryl thioacetate

    Specifications

    Formula Weight 156.2
    TSCA TSCA
    Molecular Formula C7H8O2S
    MDL Number MFCD00010083
    CAS 13678-68-7
    UN Number UN3334
    Synonym Thioacetic acid S-furfuryl ester
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 211° to 212°C
    Quantity 25g
    Flash Point 92°C (199°F)
    Odor Stench
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
    Chemical Name or Material S-Furfuryl thioacetate
    Assay 99%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit