Alfa Aesar™ S-Furfuryl thiopropionate, 99%

Manufacturer: Alfa Aesar™ B2235106

Quantity 5g

 View more versions of this product

Catalog No. AAB2235106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59020-85-8
    Molecular Formula C8H10O2S
    Formula Weight 170.23
    MDL Number MFCD00040264
    Synonym Thiopropionic acid S-furfuryl ester
    Chemical Name or Material S-Furfuryl thiopropionate

    Specifications

    Formula Weight 170.23
    Flash Point 97°C (207°F)
    TSCA TSCA
    Molecular Formula C8H10O2S
    MDL Number MFCD00040264
    CAS 59020-85-8
    Synonym Thiopropionic acid S-furfuryl ester
    Boiling Point 219° to 221°C
    Quantity 5g
    Chemical Name or Material S-Furfuryl thiopropionate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit