Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-(-)-HA-966 Available on GSA/VA Contract for Federal Government customers only.

$99.00 - $425.00

Specifications

Assay Percent Range >98%
Formula Weight Observed MW: 116.12
Chemical Name or Material (S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one
Molecular Formula C4H8N2O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
02-825-0 View Documents Tocris Bioscience™
0282/50
50mg Each for $425.00
Add to cart
 
02-821-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
0282/10
10mg Each for $99.00
Add to cart
 
Description

Description

Possesses potent sedative and ataxic action, probably through disruption of striatal dopaminergic mechanisms.

R-enantiomer also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

>98%
Observed MW: 116.12
(S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one
C4H8N2O2
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit