Learn More
Tocris Bioscience™ (S)-(-)-HA-966
NMDA antagonist/partial agonist
Manufacturer:  Tocris Bioscience™ 0282/50
 View more versions of this product
Catalog No. 02-825-0
  Check Availability
Add to cart
Description
Description
Possesses potent sedative and ataxic action, probably through disruption of striatal dopaminergic mechanisms. R-enantiomer (Cat. No. 0281) also available.
Specifications
Specifications
(S)-(-)-3-Amino-1-hydroxypyrrolidin-2-one
>98%
Observed MW: 116.12
Soluble to 100 mM in water
111821-58-0
C4H8N2O2
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT