Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Isopropylisothiourea hydrobromide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

4269-97-0
C4H11BrN2S
199.11
SLGVZEOMLCTKRK-UHFFFAOYSA-N
9813062
propan-2-yl carbamimidothioate;hydrobromide
CC(C)SC(=N)N.Br

iNOS inhibitor, acts at arginine binding site

Potent inhibitor of NO synthase, with selectivity for the inducible form.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 199.11
Chemical Name or Material S-Isopropylisothiourea hydrobromide
Grade >98%

Recommended Storage : Store at Room Temperature