Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Mandelic Acid ≥99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
17199-29-0
C8H8O3
152.149
MFCD00004495
IWYDHOAUDWTVEP-ZETCQYMHSA-N
s-+-mandelic acid,s-mandelic acid,s-2-hydroxy-2-phenylacetic acid,l-mandelic acid,l-+-mandelic acid,2s-2-hydroxy-2-phenylacetic acid,mandelic acid, s,unii-l0umw58g3t,l +-mandelic acid
439616
CHEBI:32800
(2S)-2-hydroxy-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)O
17199-29-0
C8H8O3
152.149
MFCD00004495
IWYDHOAUDWTVEP-ZETCQYMHSA-N
s-+-mandelic acid,s-mandelic acid,s-2-hydroxy-2-phenylacetic acid,l-mandelic acid,l-+-mandelic acid,2s-2-hydroxy-2-phenylacetic acid,mandelic acid, s,unii-l0umw58g3t,l +-mandelic acid
439616
CHEBI:32800
(2S)-2-hydroxy-2-phenylacetic acid
C1=CC=C(C=C1)C(C(=O)O)O
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H5CH(OH)CO2H
Beilstein 10, 192
Formula Weight 152.15
Physical Form Crystalline Powder
Percent Purity ≥99.0% (Acid)
Infrared Spectrum Authentic
Packaging Plastic bottle
Specific Rotation +152° to +158° (20°C, 589nm) (c=2, H2O)
Sulfated Ash 0.1% max.
Water 1% max. (K.F.)
Specific Rotation Condition + 155.00 (20.00°C c=2,H2O)
Melting Point 130°C to 135°C
Color Beige-Brown to White
Quantity 25g
Chemical Name or Material (S)-(+)-Mandelic acid, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LIGHT SENSITIVE

EINECSNumber : 241-240-8

TSCA : TSCA