Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ S-Methyl thiobutyrate, 98%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
2432-51-1
C5H10OS
118.194
MFCD00009872
GRLJIIJNZJVMGP-UHFFFAOYSA-N
s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
62444
S-methyl butanethioate
CCCC(=O)SC
2432-51-1
C5H10OS
118.194
MFCD00009872
GRLJIIJNZJVMGP-UHFFFAOYSA-N
s-methyl thiobutyrate, butanethioic acid, s-methyl ester, methyl thiolbutyrate, methanethiol butyrate, methanethiol n-butyrate, butyric acid, thio-, s-methyl ester, s-methyl thiobutanoate, unii-2p1e432myz, methyl thiobutanoate, methyl thiobutyrate natural
62444
S-methyl butanethioate
CCCC(=O)SC
Density 0.966
Refractive Index 1.462
Quantity 25g
Boiling Point 142°C to 143°C
Flash Point 34°C (93°F)
UN Number UN1993
Beilstein 1848987
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material S-Methyl thiobutyrate

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 219-407-1

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only