Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-α-Methylaspartic acid-4-tert-butyl ester, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
1217977-71-3
C9H17NO4
203.24
VIQZVYPYYUPDQY-VIFPVBQESA-N
s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid,s-alpha-methylaspartic acid-4-t-butyl ester,s-alpha-methylasparticacid-4-tert-butylester,s-alpha-methylaspartic acid-4-tert-butyl ester,h-alpha-me-l-asp tbu-oh,r-a-methylaspartic acid-4-tert-butyl ester,2s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid
51885231
(2S)-2-amino-2-methyl-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
CC(C)(C)OC(=O)CC(C)(C(=O)O)N
1217977-71-3
C9H17NO4
203.24
VIQZVYPYYUPDQY-VIFPVBQESA-N
s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid,s-alpha-methylaspartic acid-4-t-butyl ester,s-alpha-methylasparticacid-4-tert-butylester,s-alpha-methylaspartic acid-4-tert-butyl ester,h-alpha-me-l-asp tbu-oh,r-a-methylaspartic acid-4-tert-butyl ester,2s-2-amino-4-tert-butoxy-2-methyl-4-oxobutanoic acid
51885231
(2S)-2-amino-2-methyl-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
CC(C)(C)OC(=O)CC(C)(C(=O)O)N
Melting Point 177°C to 178°C
Specific Rotation 33
Quantity 100mg
Color White to Yellow
Infrared Spectrum Authentic
Enantiomeric Excess 98.0% min.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min
Formula Weight 203.24
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (S)-α-Methylaspartic acid-4-tert-butyl ester, 98% ee
Show More Show Less