Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-α-Methylbenzyl isocyanate, 98%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
14649-03-7
C9H9NO
147.18
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate,1s-1-isocyanatoethyl benzene,s---alpha-methylbenzyl isocyanate,benzene, 1s-1-isocyanatoethyl,isocyanic acid s---a-methylbenzyl ester,s-alpha-methylbenzyl isocyanate,s---1-phenylethylisocyanate,isocyanic acid s---alpha-methylbenzyl ester,isocyanic acid s---methylbenzyl ester,1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
14649-03-7
C9H9NO
147.18
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate,1s-1-isocyanatoethyl benzene,s---alpha-methylbenzyl isocyanate,benzene, 1s-1-isocyanatoethyl,isocyanic acid s---a-methylbenzyl ester,s-alpha-methylbenzyl isocyanate,s---1-phenylethylisocyanate,isocyanic acid s---alpha-methylbenzyl ester,isocyanic acid s---methylbenzyl ester,1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
Chiral derivatization reagent
Specific Rotation − 10.00
Specific Rotation Condition −10° (20°C neat)
Specific Gravity 1.05
Flash Point 65°C
Color Colorless to Yellow
Refractive Index 1.5130 to 1.5150
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 55.0°C to 56.0°C (2.5mmHg)
Density 1.0500g/mL
Quantity 5mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CH(CH3)NCO
Solubility Information Solubility in water: decomposes
Formula Weight 147.18
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (S)-(-)-α-Methylbenzyl isocyanate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Fatal if inhaled.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 238-698-6

TSCA : TSCA