Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Methyl methanethiolsulfonate, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
100g
Packaging:
Glass bottle
2949-92-0
C2H6O2S2
126.19
MFCD00007565
XYONNSVDNIRXKZ-UHFFFAOYSA-N
s-methyl methanethiosulfonate,methyl methanethiolsulfonate,methyl methanethiosulfonate,s-methyl methanesulfonothioate,methanesulfonothioic acid, s-methyl ester,methyl methanesulfonothioate,s-methyl methanethiolsulfonate,methanethiosulfonic acid s-methyl ester,s-methyl methanethiosulphonate,methanesulfonylsulfanyl methane
18064
CHEBI:74357
methylsulfonylsulfanylmethane
CSS(=O)(=O)C
2949-92-0
C2H6O2S2
126.19
MFCD00007565
XYONNSVDNIRXKZ-UHFFFAOYSA-N
s-methyl methanethiosulfonate,methyl methanethiolsulfonate,methyl methanethiosulfonate,s-methyl methanesulfonothioate,methanesulfonothioic acid, s-methyl ester,methyl methanesulfonothioate,s-methyl methanethiolsulfonate,methanethiosulfonic acid s-methyl ester,s-methyl methanethiosulphonate,methanesulfonylsulfanyl methane
18064
CHEBI:74357
methylsulfonylsulfanylmethane
CSS(=O)(=O)C
Specific Gravity 1.227
Boiling Point 69.0°C to 71.0°C (0.4mmHg)
Refractive Index 1.5118
Packaging Glass bottle
Quantity 1g
Density 1.2270g/mL
Flash Point 87°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3SO2SCH3
Fieser 05,454; 07,243; 12,325; 01,169; 10,107; 16,99
Formula Weight 126.19
Percent Purity 97%
Chemical Name or Material S-Methyl methanethiolsulfonate
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Warning

EINECSNumber : 220-970-