Learn More
Tocris Bioscience™ (S)-(+)-α-Methylhistamine dihydrobromide
Manufacturer:  Tocris Bioscience™ 0572/50
 View more versions of this product
Catalog No. 05-725-0
Add to cart
This item is not returnable. View return policy
Description
Description
Less active enantiomer of H3 agonist R-(-)-α-methylhistamine ; 120-fold less potent than R-(-) at H3.
R-enantiomer also available.
Specifications
Specifications
(S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
C6H11N3.2HBr
50mg
75614-93-6
Observed MW: 287
Soluble to 100 mM in water
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at RT