Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-(+)-α-Methylhistamine dihydrobromide

$565.00

Specifications

Formula Weight Observed MW: 287
Chemical Name or Material (S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
Molecular Formula C6H11N3.2HBr
CAS 75614-93-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-725-0
View Documents
Tocris Bioscience™
0572/50
50 mg
Each for $565.00
Add to cart
 
05-721-0
View Documents
Tocris Bioscience™
0572/10
10 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Less active enantiomer of H3 agonist R-(-)-α-methylhistamine ; 120-fold less potent than R-(-) at H3.

R-enantiomer also available.

Specifications

Specifications

Observed MW: 287
(S)-(+)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
C6H11N3.2HBr
75614-93-6
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at Room Temperature

SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit