Alfa Aesar™ S-Methylisothiouronium sulfate, 98+%

Manufacturer: Alfa Aesar™ A1104436

Quantity 500g

 View more versions of this product

Catalog No. AAA1104436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 867-44-7
    Molecular Formula C4H12N4S2·H2SO4
    Formula Weight 278.38
    MDL Number MFCD00013055
    Beilstein 3917217
    Synonym S-Methylisothiourea sulfate; 2-Methyl-2-thiopseudourea sulfate
    Chemical Name or Material S-Methylisothiouronium sulfate

    Specifications

    Formula Weight 278.38
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 867-44-7
    Beilstein 3917217
    Molecular Formula C4H12N4S2·H2SO4
    MDL Number MFCD00013055
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Melting Point ≈235°C (dec.)
    RTECS Number UM9200000
    TSCA TSCA
    Synonym S-Methylisothiourea sulfate; 2-Methyl-2-thiopseudourea sulfate
    Health Hazard 1 Danger
    Quantity 500g
    Chemical Name or Material S-Methylisothiouronium sulfate
    Assay ≥98%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit