Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-α-Methylleucine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
105743-53-1
C7H15NO2
145.2
ARSWQPLPYROOBG-ZETCQYMHSA-N
2-methylleucine,l-leucine, 2-methyl,s-2-amino-2,4-dimethylpentanoic acid,h-alpha-me-leu-oh,h-alpha-me-l-leu-oh,2s-2-amino-2,4-dimethylpentanoic acid,s-alpha-methylleucine,2ml,a-methylleucine,ambotzhaa5400
446181
(2S)-2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
105743-53-1
C7H15NO2
145.2
ARSWQPLPYROOBG-ZETCQYMHSA-N
2-methylleucine,l-leucine, 2-methyl,s-2-amino-2,4-dimethylpentanoic acid,h-alpha-me-leu-oh,h-alpha-me-l-leu-oh,2s-2-amino-2,4-dimethylpentanoic acid,s-alpha-methylleucine,2ml,a-methylleucine,ambotzhaa5400
446181
(2S)-2-amino-2,4-dimethylpentanoic acid
CC(C)CC(C)(C(=O)O)N
Specific Rotation 35
Melting Point >237°C
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Color White to Yellow
Quantity 100mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5 to 102.5%
Formula Weight 145.2
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (S)-α-Methylleucine, 98% ee
Show More Show Less