Learn More
Medchemexpress LLC (S)-Minzasolmin | 1802518-91-7 | 99.4% | 425.59 g/mol | C23H31N5OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145580A5MG
Encompass_Preferred
(S)-Minzasolmin is the S-enantiomer of minzasolmin, a small-molecule inhibitor of α-synuclein oligomerization supplied for research use. It is intended for preclinical biochemical and pharmacological studies investigating synuclein aggregation and related disease models, and is provided as a characterized, high-purity reagent.
  • High purity suitable for analytical and biological assays.
  • Chiral S-enantiomer for stereospecific activity studies.
  • Active in inhibiting α-synuclein oligomerization in vitro and in vivo models.
  • Provided in small milligram quantities for research workflows.
  • Characterized by formula C23H31N5OS and molecular weight 425.59 g/mol.
Catalog No. 50-002-21073