Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
66849-29-4
C10H15NO
165.23
MFCD01862172
GXIWMXAAPLZOBY-VIFPVBQESA-N
s-2-1-phenylethyl amino ethanol,2-1s-1-phenylethyl amino ethan-1-ol,2-1s-1-phenylethyl amino ethanol,s---n-2-hydroxyethyl-alpha-phenylethylamine,2-s-alpha-methylbenzyl amino ethanol,s-n-2-hydroxyethyl-a-phenylethylamine,ethanol,2-1s-1-phenylethyl amino,s-+-n-2-hydroxyethyl-phenylethylamine,s---n-2-hydroxyethyl-alpha-methylbenzylamine
2733847
2-[[(1S)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
66849-29-4
C10H15NO
165.23
MFCD01862172
GXIWMXAAPLZOBY-VIFPVBQESA-N
s-2-1-phenylethyl amino ethanol,2-1s-1-phenylethyl amino ethan-1-ol,2-1s-1-phenylethyl amino ethanol,s---n-2-hydroxyethyl-alpha-phenylethylamine,2-s-alpha-methylbenzyl amino ethanol,s-n-2-hydroxyethyl-a-phenylethylamine,ethanol,2-1s-1-phenylethyl amino,s-+-n-2-hydroxyethyl-phenylethylamine,s---n-2-hydroxyethyl-alpha-methylbenzylamine
2733847
2-[[(1S)-1-phenylethyl]amino]ethanol
CC(C1=CC=CC=C1)NCCO
Specific Rotation Condition − 44.00 (22.00°C c=1,C2H5OH)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5350
Infrared Spectrum Authentic
Specific Rotation − 44.00
Color Colorless
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH(CH3)NH(CH2)2OH
Formula Weight 165.23
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA