Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 95%

Click to view available options
Quantity:
5g
25g
79261-58-8
C18H17NO4
311.337
MFCD00672545
YWVQGUBCAUFBCP-INIZCTEOSA-N
3s-2-carbobenzoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,z-tic-oh,s-2-benzyloxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,s-n-cbz-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,s-+-2-benzyloxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid,z-3s-1,2,3,4-tetrahydroisoquinolene-3-carboxylic acid,3s-2-carbobenzoxy-1,2,3,4-tetrahydroisoquinoline,3s-2-benzyloxy carbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid,s-+-n-cbz-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,3s-2-benzyloxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
688307
(3S)-2-phenylmethoxycarbonyl-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
C1C(N(CC2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)C(=O)O
79261-58-8
C18H17NO4
311.337
MFCD00672545
YWVQGUBCAUFBCP-INIZCTEOSA-N
3s-2-carbobenzoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,z-tic-oh,s-2-benzyloxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,s-n-cbz-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,s-+-2-benzyloxycarbonyl-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid,z-3s-1,2,3,4-tetrahydroisoquinolene-3-carboxylic acid,3s-2-carbobenzoxy-1,2,3,4-tetrahydroisoquinoline,3s-2-benzyloxy carbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid,s-+-n-cbz-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,3s-2-benzyloxy carbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
688307
(3S)-2-phenylmethoxycarbonyl-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
C1C(N(CC2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)C(=O)O
Color White to Yellow
Melting Point 136°C to 140°C
Quantity 25g
Beilstein 7221387
Formula Weight 311.34
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (S)-N-Benzyloxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

storageNote1 : 2° to 8°C