Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-1-Naphthylalanine, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
55447-00-2
C18H21NO4
315.369
MFCD00085377
KHHIGWRTNILXLL-HNNXBMFYSA-N
boc-3-1-naphthyl-l-alanine,boc-l-1-naphthylalanine,boc-1-nal-oh,s-n-boc-l-1-naphthyl alanine,boc-l-3-1-naphthyl-alanine,boc-d-1-nal-oh,boc-3-1-naphthyl-l-ala,2s-2-tert-butoxy carbonylamino-3-naphthylpropanoic acid,3-naphthalen-1-yl-n-boc-l-alanine,boc-d-3-1-naphthyl-alanine
7021841
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-1-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC2=CC=CC=C21)C(=O)O
55447-00-2
C18H21NO4
315.369
MFCD00085377
KHHIGWRTNILXLL-HNNXBMFYSA-N
boc-3-1-naphthyl-l-alanine,boc-l-1-naphthylalanine,boc-1-nal-oh,s-n-boc-l-1-naphthyl alanine,boc-l-3-1-naphthyl-alanine,boc-d-1-nal-oh,boc-3-1-naphthyl-l-ala,2s-2-tert-butoxy carbonylamino-3-naphthylpropanoic acid,3-naphthalen-1-yl-n-boc-l-alanine,boc-d-3-1-naphthyl-alanine
7021841
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-1-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC2=CC=CC=C21)C(=O)O
Color White to Beige
Packaging Glass bottle
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 1g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 315.37
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-1-Naphthylalanine, 95%, (98% e.e.)
Show More Show Less