Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
261165-06-4
C14H18BrNO4
344.205
MFCD03419292
ZAMLGGRVTAXBHI-NSHDSACASA-N
s-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid,boc-s-3-amino-3-4-bromo-phenyl-propionic acid,s-3-boc-amino-3-4-bromophenyl-propanoic acid,s-boc-4-bromo-beta-phe-oh,s-3-boc-amino-3-4-bromophenyl propionic acid,3s-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid,pubchem17339,boc-?-phe 4-br-oh,boc-r-3-amino-3-4-bromophenyl-propionic acid,boc-4-bromo-d-,a-phenylalanine
2734491
(3S)-3-(4-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=C(C=C1)Br
261165-06-4
C14H18BrNO4
344.205
MFCD03419292
ZAMLGGRVTAXBHI-NSHDSACASA-N
s-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid,boc-s-3-amino-3-4-bromo-phenyl-propionic acid,s-3-boc-amino-3-4-bromophenyl-propanoic acid,s-boc-4-bromo-beta-phe-oh,s-3-boc-amino-3-4-bromophenyl propionic acid,3s-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid,pubchem17339,boc-?-phe 4-br-oh,boc-r-3-amino-3-4-bromophenyl-propionic acid,boc-4-bromo-d-,a-phenylalanine
2734491
(3S)-3-(4-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=C(C=C1)Br
Packaging Glass bottle
Enantiomeric Excess 97.5% min.
Color White
Quantity 500mg
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 344.21
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, (98% e.e.)
Show More Show Less