Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
103365-47-5
C14H19NO4
265.309
MFCD01860892
JTNQFJPZRTURSI-NSHDSACASA-N
boc-s-3-amino-3-phenylpropionic acid,s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,s-3-n-boc-amino-beta-phenylalanine,s-n-boc-3-amino-3-phenylpropanoic acid,boc-d-beta-phe-oh,boc-d-beta-phenylalanine,s-boc-beta3-phenylalanine,boc-s-phenylalanine,3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,s-3-boc-amino-3-phenylpropionic acid
2734490
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
103365-47-5
C14H19NO4
265.309
MFCD01860892
JTNQFJPZRTURSI-NSHDSACASA-N
boc-s-3-amino-3-phenylpropionic acid,s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,s-3-n-boc-amino-beta-phenylalanine,s-n-boc-3-amino-3-phenylpropanoic acid,boc-d-beta-phe-oh,boc-d-beta-phenylalanine,s-boc-beta3-phenylalanine,boc-s-phenylalanine,3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid,s-3-boc-amino-3-phenylpropionic acid
2734490
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1
Color White
Enantiomeric Excess 97.5% min.
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 265.31
Physical Form Chunks
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-3-Amino-3-phenylpropanoic acid, 95%, (98% e.e.)
Show More Show Less