Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-3-Cyanophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
131980-30-8
C15H18N2O4
290.319
MFCD00797560
FDQDHMZKOPOWFE-LBPRGKRZSA-N
boc-phe 3-cn-oh,boc-l-3-cyanophenylalanine,boc-3-cyano-l-phenylalanine,s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid,boc-l-3-cn-phe-oh,2s-2-tert-butoxy carbonylamino-3-3-cyanophenyl propanoic acid,boc-l-3-cyanophe,boc-l-3-cyano phenylalanine
2734498
(2S)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O
131980-30-8
C15H18N2O4
290.319
MFCD00797560
FDQDHMZKOPOWFE-LBPRGKRZSA-N
boc-phe 3-cn-oh,boc-l-3-cyanophenylalanine,boc-3-cyano-l-phenylalanine,s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid,2s-2-tert-butoxycarbonyl amino-3-3-cyanophenyl propanoic acid,boc-l-3-cn-phe-oh,2s-2-tert-butoxy carbonylamino-3-3-cyanophenyl propanoic acid,boc-l-3-cyanophe,boc-l-3-cyano phenylalanine
2734498
(2S)-3-(3-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=CC(=C1)C#N)C(=O)O
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White to Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 290.31
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-3-Cyanophenylalanine, 95%, (98% e.e.)
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA