Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-(3-Pyridyl)alanine, 95%, 98% ee, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
117142-26-4
C13H18N2O4
266.29
MFCD00079681
JLBCSWWZSSVXRQ-JTQLQIEISA-N
boc-3-3-pyridyl-l-alanine,boc-3-pal-oh,boc-3-3-pyridyl-alanine,boc-l-3-3-pyridyl-alanine,s-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid,boc-3-3-pyridyl-ala-oh,boc-ala 3-pyridyl-oh,boc-l-3-pyridylala,boc-3'-pyridyl-l-ala,n-tert-butoxycarbonyl-3-3-pyridyl-l-alanine
7020919
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O
117142-26-4
C13H18N2O4
266.29
MFCD00079681
JLBCSWWZSSVXRQ-JTQLQIEISA-N
boc-3-3-pyridyl-l-alanine,boc-3-pal-oh,boc-3-3-pyridyl-alanine,boc-l-3-3-pyridyl-alanine,s-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid,boc-3-3-pyridyl-ala-oh,boc-ala 3-pyridyl-oh,boc-l-3-pyridylala,boc-3'-pyridyl-l-ala,n-tert-butoxycarbonyl-3-3-pyridyl-l-alanine
7020919
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O
Melting Point 135°C to 138°C
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 266.29
Physical Form Crystalline Powder
Percent Purity 95%,98% ee
Chemical Name or Material (S)-N-BOC-(3-Pyridyl)alanine, 95%, (98% e.e.)
Show More Show Less