Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
68090-88-0
C14H18ClNO4
299.751
MFCD00057808
BETBOAZCLSJOBQ-NSHDSACASA-N
boc-phe 4-cl-oh,boc-l-4-chlorophenylalanine,boc-4-chloro-l-phenylalanine,boc-l-4-chlorophe,boc-l-phe 4-cl-oh,n-boc-4-chloro-l-phenylalanine,n-tert-butoxycarbonyl-4-chloro-l-phenylalanine,s-n-boc-4-chlorophenylalanine,s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid,boc-p-chloro-phe-oh
2734499
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Cl)C(=O)O
68090-88-0
C14H18ClNO4
299.751
MFCD00057808
BETBOAZCLSJOBQ-NSHDSACASA-N
boc-phe 4-cl-oh,boc-l-4-chlorophenylalanine,boc-4-chloro-l-phenylalanine,boc-l-4-chlorophe,boc-l-phe 4-cl-oh,n-boc-4-chloro-l-phenylalanine,n-tert-butoxycarbonyl-4-chloro-l-phenylalanine,s-n-boc-4-chlorophenylalanine,s-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid,boc-p-chloro-phe-oh
2734499
(2S)-3-(4-chlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Cl)C(=O)O
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color Yellow
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 299.75
Physical Form Granular Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-4-Chlorophenylalanine, 95%, (98% e.e.)
Show More Show Less