Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-4-Fluorophenylalanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
41153-30-4
C14H18FNO4
283.299
MFCD00079672
RCXSXRAUMLKRRL-NSHDSACASA-N
boc-l-4-fluorophenylalanine,boc-phe 4-f-oh,boc-l-4-fluorophe,boc-4-fluoro-l-phenylalanine,boc-4-fluoro-l-phe-oh,n-tert-butoxycarbonyl-4-fluoro-l-phenylalanine,s-n-boc-4-fluorophenylalanine,boc-l-phe 4-f-oh,boc-p-fluoro-l-phenylalanine,boc-p-fluoro-phe-oh
7020910
(2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O
41153-30-4
C14H18FNO4
283.299
MFCD00079672
RCXSXRAUMLKRRL-NSHDSACASA-N
boc-l-4-fluorophenylalanine,boc-phe 4-f-oh,boc-l-4-fluorophe,boc-4-fluoro-l-phenylalanine,boc-4-fluoro-l-phe-oh,n-tert-butoxycarbonyl-4-fluoro-l-phenylalanine,s-n-boc-4-fluorophenylalanine,boc-l-phe 4-f-oh,boc-p-fluoro-l-phenylalanine,boc-p-fluoro-phe-oh
7020910
(2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O
Infrared Spectrum Authentic
Quantity 1g
Enantiomeric Excess 97.5% min.
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 283.3
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-4-Fluorophenylalanine, 95%, (98% e.e.)
Show More Show Less