Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™
Click to view available options
Quantity:
500mg
1g
Specifications
Quantity | 1g |
Color | White |
Melting Point | 130.2°C |
Infrared Spectrum | Authentic |
Enantiomeric Excess | 97.5% min. (HPLC) |
Packaging | Glass bottle |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Formula Weight | 269.27 |
Show More |
Safety and Handling
TSCA : TSCA