Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-4-Fluorophenylglycine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
142186-36-5
C13H16FNO4
269.272
MFCD01320862
IEQBOUSLFRLKKX-JTQLQIEISA-N
s-2-tert-butoxycarbonyl amino-2-4-fluorophenyl acetic acid,s-n-boc-4-fluorophenylglycine,s-tert-butoxycarbonyl amino 4-fluorophenyl acetic acid,2s-tert-butoxycarbonyl amino 4-fluorophenyl ethanoic acid,boc-4-fluoro-l-phenylglycine,n-tert-butoxycarbonyl-4-fluoro-l-phenylglycine,2s-tert-butoxycarbonyl amino 4-fluorophenyl acetic acid,l-n-tert-butoxycarbonyl-alpha-4-fluorophenyl acetic acid,s-2-tert-butoxycarbonyl amino-2-4-fluorophenyl aceticacid,s-tert-butoxycarbonylamino-4-fluoro-phenyl-acetic acid
2734497
(2S)-2-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1=CC=C(C=C1)F)C(=O)O
142186-36-5
C13H16FNO4
269.272
MFCD01320862
IEQBOUSLFRLKKX-JTQLQIEISA-N
s-2-tert-butoxycarbonyl amino-2-4-fluorophenyl acetic acid,s-n-boc-4-fluorophenylglycine,s-tert-butoxycarbonyl amino 4-fluorophenyl acetic acid,2s-tert-butoxycarbonyl amino 4-fluorophenyl ethanoic acid,boc-4-fluoro-l-phenylglycine,n-tert-butoxycarbonyl-4-fluoro-l-phenylglycine,2s-tert-butoxycarbonyl amino 4-fluorophenyl acetic acid,l-n-tert-butoxycarbonyl-alpha-4-fluorophenyl acetic acid,s-2-tert-butoxycarbonyl amino-2-4-fluorophenyl aceticacid,s-tert-butoxycarbonylamino-4-fluoro-phenyl-acetic acid
2734497
(2S)-2-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetic acid
CC(C)(C)OC(=O)NC(C1=CC=C(C=C1)F)C(=O)O
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (HPLC)
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 269.27
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-4-Fluorophenylglycine, 95%, (98% e.e.)
Show More Show Less