Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-Allylglycine, 95%, ACROS Organics™

Click to view available options
Quantity:
500mg
1g
5g
Packaging:
Glass bottle
90600-20-7
C10H17NO4
215.249
MFCD01320851
BUPDPLXLAKNJMI-ZETCQYMHSA-N
s-2-tert-butoxycarbonyl amino pent-4-enoic acid,s-n-boc-allylglycine,boc-l-allylglycine,boc-alpha-allyl-l-gly,4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s,s-2-tert-butoxycarbonylamino pent-4-enoic acid,2s-2-tert-butoxycarbonyl amino pent-4-enoic acid,boc-l-alpha-allyl-gly,boc--allyl-l-gly,rarechem bk pt 0250
2734487
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
90600-20-7
C10H17NO4
215.249
MFCD01320851
BUPDPLXLAKNJMI-ZETCQYMHSA-N
s-2-tert-butoxycarbonyl amino pent-4-enoic acid,s-n-boc-allylglycine,boc-l-allylglycine,boc-alpha-allyl-l-gly,4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s,s-2-tert-butoxycarbonylamino pent-4-enoic acid,2s-2-tert-butoxycarbonyl amino pent-4-enoic acid,boc-l-alpha-allyl-gly,boc--allyl-l-gly,rarechem bk pt 0250
2734487
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 215.25
Specific Rotation + 10.50
Physical Form Viscous Liquid
Percent Purity 95%
Specific Rotation Condition +10.50° (20°C c=1,methanol)
Color Yellow
Quantity 1g
Chemical Name or Material (S)-N-BOC-Allylglycine
Show More Show Less