Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-Azetidine-2-carboxylic acid, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
51077-14-6
C9H15NO4
201.222
MFCD01321011
JWJVSDZKYYXDDN-LURJTMIESA-N
s-1-tert-butoxycarbonyl azetidine-2-carboxylic acid,s-n-boc-azetidine carboxylic acid,1-boc-l-azetidine-2-carboxylic acid,s-1-boc-azetidine-2-carboxylic acid,boc-aze-oh,s-n-boc-azetidine-2-carboxylic acid,boc-aze 2-oh,boc-s-azetidine-2-carboxylic acid,boc-l-azetidine-2-carboxylic acid,2s-1-tert-butoxycarbonyl azetidine-2-carboxylic acid
2734495
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]azetidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCC1C(=O)O
51077-14-6
C9H15NO4
201.222
MFCD01321011
JWJVSDZKYYXDDN-LURJTMIESA-N
s-1-tert-butoxycarbonyl azetidine-2-carboxylic acid,s-n-boc-azetidine carboxylic acid,1-boc-l-azetidine-2-carboxylic acid,s-1-boc-azetidine-2-carboxylic acid,boc-aze-oh,s-n-boc-azetidine-2-carboxylic acid,boc-aze 2-oh,boc-s-azetidine-2-carboxylic acid,boc-l-azetidine-2-carboxylic acid,2s-1-tert-butoxycarbonyl azetidine-2-carboxylic acid
2734495
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]azetidine-2-carboxylic acid
CC(C)(C)OC(=O)N1CCC1C(=O)O
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 500mg
Infrared Spectrum Authentic
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 201.22
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-BOC-Azetidine-2-carboxylic acid, 95%, (98% e.e.)
Show More Show Less