Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-α-Ethylalanine, 98%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
151171-11-8
C10H19NO4
217.27
SHZXLTCEPXVCSV-JTQLQIEISA-N
boc-l-isovaline,s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid,s-2-tert-butoxycarbonylamino-2-methylbutanoic acid,n-1,1-dimethylethyl oxy carbonyl-d-isovaline,s-2-methyl-2-tert-butoxycarbonylamino butyric acid,2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid,2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
14284792
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C)(C(=O)O)NC(=O)OC(C)(C)C
151171-11-8
C10H19NO4
217.27
SHZXLTCEPXVCSV-JTQLQIEISA-N
boc-l-isovaline,s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid,s-2-tert-butoxycarbonylamino-2-methylbutanoic acid,n-1,1-dimethylethyl oxy carbonyl-d-isovaline,s-2-methyl-2-tert-butoxycarbonylamino butyric acid,2s-2-tert-butoxy carbonyl amino-2-methylbutanoic acid,2s-2-tert-butoxycarbonyl amino-2-methylbutanoic acid
14284792
(2S)-2-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
CCC(C)(C(=O)O)NC(=O)OC(C)(C)C
Specific Rotation 12
Melting Point 104°C to 112°C
Packaging Glass Bottle
Quantity 500mg
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 217.27
Physical Form Powder
Percent Purity 98%,98% ee
Chemical Name or Material (S)-N-BOC-α-Ethylalanine, 98% ee
Show More Show Less