Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-BOC-Prolinal, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
69610-41-9
C10H17NO3
199.25
MFCD00274186
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal,boc-l-prolinal,s-1-boc-2-formylpyrrolidine,s-tert-butyl 2-formylpyrrolidine-1-carboxylate,tert-butyl 2s-2-formylpyrrolidine-1-carboxylate,boc-prolinal,n-tert-butoxycarbonyl-l-prolinal,s-n-tert-butoxycarbonyl-2-formylpyrrolidine,1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s,n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1C=O
69610-41-9
C10H17NO3
199.25
MFCD00274186
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal,boc-l-prolinal,s-1-boc-2-formylpyrrolidine,s-tert-butyl 2-formylpyrrolidine-1-carboxylate,tert-butyl 2s-2-formylpyrrolidine-1-carboxylate,boc-prolinal,n-tert-butoxycarbonyl-l-prolinal,s-n-tert-butoxycarbonyl-2-formylpyrrolidine,1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s,n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC1C=O
Quantity 5g
Density 1.0630g/mL
Boiling Point 211.0°C
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630
Specific Rotation − 97.50
Specific Gravity 1.063
Specific Rotation Condition −97.5° (20°C c=1,CHCl3)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material (S)-N-BOC-Prolinal, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Freezer -20°C