Alfa Aesar™ (S)-N-Diphenylphosphino-N-methyl-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™

Asymmetric hydrogenation

Manufacturer: Alfa Aesar™ 44685-02

Quantity 0.5g

 View more versions of this product

Catalog No. AA4468502

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 406681-09-2
    Molecular Formula C37H35FeNP2
    Formula Weight 611.5
    MDL Number MFCD05865220
    Synonym (S)-Methyl BoPhoz™
    Chemical Name or Material (S)-N-Diphenylphosphino-N-methyl-[ (R)-2- (diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™

    Specifications

    Formula Weight 611.5
    Synonym (S)-Methyl BoPhoz™
    Physical Form Solid
    Molecular Formula C37H35FeNP2
    MDL Number MFCD05865220
    Color Yellow to orange
    Odor Odorless
    CAS 406681-09-2
    Melting Point 80° to 82°C
    Quantity 0.5g
    Chemical Name or Material (S)-N-Diphenylphosphino-N-methyl-[ (R)-2- (diphenylphosphino)ferrocenyl]ethylamine, (S)-Methyl BoPhoz™

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit