Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-1-Naphthylalanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
96402-49-2
C28H23NO4
437.495
MFCD00151914
ORWNVJDLEMVDLV-SANMLTNESA-N
fmoc-3-1-naphthyl-l-alanine,fmoc-1-nal-oh,fmoc-l-1-naphthylalanine,fmoc-beta-1-naphthyl-l-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,fmoc-3-1 naphthyl-l-alanine,fmoc-l-3-1-naphthyl-alanine,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,n-fmoc-3-1-naphthyl-l-alanine
2734474
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-1-ylpropanoic acid
C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
96402-49-2
C28H23NO4
437.495
MFCD00151914
ORWNVJDLEMVDLV-SANMLTNESA-N
fmoc-3-1-naphthyl-l-alanine,fmoc-1-nal-oh,fmoc-l-1-naphthylalanine,fmoc-beta-1-naphthyl-l-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,fmoc-3-1 naphthyl-l-alanine,fmoc-l-3-1-naphthyl-alanine,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid,n-fmoc-3-1-naphthyl-l-alanine
2734474
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-1-ylpropanoic acid
C1=CC=C2C(=C1)C=CC=C2CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Enantiomeric Excess 97.5% min. (HPLC)
Color White to Beige
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 437.5
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-1-Naphthylalanine, 95%, (98% e.e.)
Show More Show Less