Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-(2-Bromoallyl)glycine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
220497-60-9
C20H18BrNO4
416.271
MFCD01311750
BYQROHMHEGJMPD-SFHVURJKSA-N
fmoc-l-2-amino-4-bromo-4-pentenoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-bromopent-4-enoic acid,s-n-fmoc-2-bromoallyl-glycine,2s-4-bromo-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid,s-4-bromo-2-9h-fluoren-9-ylmethoxycarbonylamino-pent-4-enoic acid,4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-pentenoic acid,4-pentenoic acid, 4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino,2s-4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-pentenoic acid,2s-4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid
2734458
(2S)-4-bromo-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
C=C(CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)Br
220497-60-9
C20H18BrNO4
416.271
MFCD01311750
BYQROHMHEGJMPD-SFHVURJKSA-N
fmoc-l-2-amino-4-bromo-4-pentenoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-bromopent-4-enoic acid,s-n-fmoc-2-bromoallyl-glycine,2s-4-bromo-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-enoic acid,s-4-bromo-2-9h-fluoren-9-ylmethoxycarbonylamino-pent-4-enoic acid,4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-pentenoic acid,4-pentenoic acid, 4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino,2s-4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-pentenoic acid,2s-4-bromo-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-enoic acid
2734458
(2S)-4-bromo-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-enoic acid
C=C(CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)Br
Color White
Packaging Glass bottle
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 500mg
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 416.27
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-(2-Bromoallyl)glycine, 95%, (98% e.e.)
Show More Show Less