Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-2-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
220497-47-2
C24H20BrNO4
466.331
MFCD01311743
GRJPAUULVKPBHU-QFIPXVFZSA-N
fmoc-l-2-bromophenylalanine,fmoc-phe 2-br-oh,fmoc-2-bromo-l-phenylalanine,s-n-fmoc-2-bromophenylalanine,fmoc-l-2-bromophe,fmoc-d-2-bromophenylalanine,fmoc-l-2-br-phe-oh,2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-bromophenyl propanoic acid,2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2734447
(2S)-3-(2-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Br
220497-47-2
C24H20BrNO4
466.331
MFCD01311743
GRJPAUULVKPBHU-QFIPXVFZSA-N
fmoc-l-2-bromophenylalanine,fmoc-phe 2-br-oh,fmoc-2-bromo-l-phenylalanine,s-n-fmoc-2-bromophenylalanine,fmoc-l-2-bromophe,fmoc-d-2-bromophenylalanine,fmoc-l-2-br-phe-oh,2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-2-bromophenyl propanoic acid,2s-3-2-bromophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2734447
(2S)-3-(2-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C(C(=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)Br
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 500mg
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 466.32
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-2-Bromophenylalanine, 95%, (98% e.e.)
Show More Show Less