Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-(2-Furyl)alanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
159611-02-6
C22H19NO5
377.396
AJXDCHXGNUFBRC-FQEVSTJZSA-N
fmoc-l-2-furylalanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid,fmoc-beta-2-furyl-ala-oh,fmoc-3-2-furyl-l-alanine,fmoc-l-3-2-furyl-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid,rarechem bk pt 0223,s-n-fmoc-furylalanine,fmoc-3-ala 2-furyl-oh
2734451
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O
159611-02-6
C22H19NO5
377.396
AJXDCHXGNUFBRC-FQEVSTJZSA-N
fmoc-l-2-furylalanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-furan-2-yl propanoic acid,fmoc-beta-2-furyl-ala-oh,fmoc-3-2-furyl-l-alanine,fmoc-l-3-2-furyl-alanine,n-9h-fluoren-9-ylmethoxy carbonyl-3-2-furyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-furan-2-yl propanoic acid,rarechem bk pt 0223,s-n-fmoc-furylalanine,fmoc-3-ala 2-furyl-oh
2734451
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(furan-2-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CO4)C(=O)O
Infrared Spectrum Authentic
Color White
Quantity 1g
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 377.39
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-(2-Furyl)alanine, 95%, (98% e.e.)
Show More Show Less