Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-2-Naphthylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
112883-43-9
C28H23NO4
437.495
MFCD00144886
JYUTZJVERLGMQZ-SANMLTNESA-N
fmoc-2-nal-oh,fmoc-3-2-naphthyl-l-alanine,fmoc-l-3-2-naphthyl-alanine,n-fmoc-3-2-naphthyl-l-alanine,fmoc-beta-2-naphthyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,s-n-fmoc-3-2-naphthyl alanine,ambotzfaa1347
2734452
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
112883-43-9
C28H23NO4
437.495
MFCD00144886
JYUTZJVERLGMQZ-SANMLTNESA-N
fmoc-2-nal-oh,fmoc-3-2-naphthyl-l-alanine,fmoc-l-3-2-naphthyl-alanine,n-fmoc-3-2-naphthyl-l-alanine,fmoc-beta-2-naphthyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid,s-n-fmoc-3-2-naphthyl alanine,ambotzfaa1347
2734452
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
C1=CC=C2C=C(C=CC2=C1)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 1g
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 437.39
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-2-Naphthylalanine, 95%, (98% e.e.)
Show More Show Less