Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-2-Thienylalanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
130309-35-2
C22H19NO4S
393.457
MFCD00065676
PXBMQFMUHRNKTG-FQEVSTJZSA-N
fmoc-l-2-thienylalanine,fmoc-3-ala 2-thienyl-oh,fmoc-l-thienylalanine,fmoc-beta-2-thienyl-ala-oh,fmoc-l-3-2-thienyl-l-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-3-2-thienyl-l-alanine,fmoc-2-thi-oh,r-n-fmoc-3-thienylalanine
7017916
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CS4)C(=O)O
130309-35-2
C22H19NO4S
393.457
MFCD00065676
PXBMQFMUHRNKTG-FQEVSTJZSA-N
fmoc-l-2-thienylalanine,fmoc-3-ala 2-thienyl-oh,fmoc-l-thienylalanine,fmoc-beta-2-thienyl-ala-oh,fmoc-l-3-2-thienyl-l-alanine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-2-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-2-yl propanoic acid,fmoc-3-2-thienyl-l-alanine,fmoc-2-thi-oh,r-n-fmoc-3-thienylalanine
7017916
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-2-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CS4)C(=O)O
Infrared Spectrum Authentic
Quantity 1g
Enantiomeric Excess 97.5% min. (HPLC)
Color White to Beige
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 393.46
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-2-Thienylalanine, 95%, (98% e.e.)
Show More Show Less