Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
500mg
1g
Packaging:
Glass bottle
220497-68-7
C24H20BrNO4
466.331
MFCD01311760
GVCLAQFMSVKNKH-QFIPXVFZSA-N
fmoc-s-3-amino-3-4-bromo-phenyl-propionic acid,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-4-bromophenyl propanoic acid,s-n-fmoc-3-amino-3-4-bromophenyl propanoic acid,pubchem17340,fmoc-?-phe 4-br-oh,fmoc-r-3-amino-3-4-bromophenyl-propionic acid,fmoc-beta-phe 4-br-oh,benzenepropanoic acid, 4-bromo-beta-9h-fluoren-9-ylmethoxy carbonyl amino-, betas,s-fmoc-4-bromo-beta-phenylalanine
2734464
(3S)-3-(4-bromophenyl)-3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C4=CC=C(C=C4)Br
220497-68-7
C24H20BrNO4
466.331
MFCD01311760
GVCLAQFMSVKNKH-QFIPXVFZSA-N
fmoc-s-3-amino-3-4-bromo-phenyl-propionic acid,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-4-bromophenyl propanoic acid,s-n-fmoc-3-amino-3-4-bromophenyl propanoic acid,pubchem17340,fmoc-?-phe 4-br-oh,fmoc-r-3-amino-3-4-bromophenyl-propionic acid,fmoc-beta-phe 4-br-oh,benzenepropanoic acid, 4-bromo-beta-9h-fluoren-9-ylmethoxy carbonyl amino-, betas,s-fmoc-4-bromo-beta-phenylalanine
2734464
(3S)-3-(4-bromophenyl)-3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC(=O)O)C4=CC=C(C=C4)Br
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 466.33
Physical Form Crystals
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-3-Amino-3-(4-bromophenyl)propanoic acid, 95%, (98% e.e.)
Show More Show Less