Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-3-Amino-3-phenylpropanoic acid, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
1g
209252-15-3
C24H21NO4
387.435
MFCD01311759
PTSLRPMRTOVHAB-QFIPXVFZSA-N
s-n-fmoc-3-amino-3-phenylpropanoic acid,fmoc-beta-s-3-phenylalanine,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-beta-d-phe-oh,fmoc-s-phenylalanine,3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid,fmoc-d-beta-phenylalanine,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,s-n-fmoc-3-amino-3-phenylpropanoicacid,fmoc-?-phe-oh
7006554
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
209252-15-3
C24H21NO4
387.435
MFCD01311759
PTSLRPMRTOVHAB-QFIPXVFZSA-N
s-n-fmoc-3-amino-3-phenylpropanoic acid,fmoc-beta-s-3-phenylalanine,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,fmoc-beta-d-phe-oh,fmoc-s-phenylalanine,3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoic acid,fmoc-d-beta-phenylalanine,s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid,s-n-fmoc-3-amino-3-phenylpropanoicacid,fmoc-?-phe-oh
7006554
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
C1=CC=C(C=C1)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Enantiomeric Excess 97.5% min.
Quantity 1g
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 387.44
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-3-Amino-3-phenylpropanoic acid, 95%, (98% e.e.)
Show More Show Less