Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-3-Bromophenylalanine, 95%, 98% e.e., Acros Organics

Click to view available options
Quantity:
500mg
220497-48-3
C24H20BrNO4
466.331
MFCD01311744
CALGTIKYXVBTAO-QFIPXVFZSA-N
fmoc-3-bromo-l-phenylalanine,fmoc-phe 3-br-oh,s-n-fmoc-3-bromophenylalanine,fmoc-l-3-bromophenylalanine,fmoc-l-3-bromophe,fmoc-l-3-br-phe-oh,2s-3-3-bromophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2s-3-3-bromophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-3-bromo-l-phe-oh,fmoc-l-phe 3-br-oh
2734448
(2S)-3-(3-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Br)C(=O)O
220497-48-3
C24H20BrNO4
466.331
MFCD01311744
CALGTIKYXVBTAO-QFIPXVFZSA-N
fmoc-3-bromo-l-phenylalanine,fmoc-phe 3-br-oh,s-n-fmoc-3-bromophenylalanine,fmoc-l-3-bromophenylalanine,fmoc-l-3-bromophe,fmoc-l-3-br-phe-oh,2s-3-3-bromophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,2s-3-3-bromophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-3-bromo-l-phe-oh,fmoc-l-phe 3-br-oh
2734448
(2S)-3-(3-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Br)C(=O)O
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Color White
Quantity 500mg
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 466.32
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-3-Bromophenylalanine, 95%, (98% e.e.)
Show More Show Less