Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-3-Thienylalanine, 95%, 98% ee, ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
186320-06-9
C22H19NO4S
393.457
MFCD00151916
LSBZJMRHROCYGY-FQEVSTJZSA-N
fmoc-l-3-thienylalanine,fmoc-3-ala 3-thienyl-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-beta-3-thienyl-ala-oh,fmoc-3-3-thienyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-d-3-3-thienyl-alanine,fmoc-l-3-3-thienyl alanine,n-fmoc-3-4-thienyl-l-alanine,fmoc-3 thg-oh
2734455
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O
186320-06-9
C22H19NO4S
393.457
MFCD00151916
LSBZJMRHROCYGY-FQEVSTJZSA-N
fmoc-l-3-thienylalanine,fmoc-3-ala 3-thienyl-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-beta-3-thienyl-ala-oh,fmoc-3-3-thienyl-l-alanine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid,fmoc-d-3-3-thienyl-alanine,fmoc-l-3-3-thienyl alanine,n-fmoc-3-4-thienyl-l-alanine,fmoc-3 thg-oh
2734455
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O
Enantiomeric Excess 97.5% min. (HPLC)
Quantity 500mg
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 393.46
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-3-Thienylalanine, 95%, (98% e.e.)
Show More Show Less