Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-4-Bromophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
198561-04-5
C24H20BrNO4
466.331
MFCD00273460
TVBAVBWXRDHONF-QFIPXVFZSA-N
fmoc-l-4-bromophenylalanine,s-n-fmoc-4-bromophenylalanine,fmoc-phe 4-br-oh,fmoc-4-bromo-l-phenylalanine,fmoc-l-4-br-phe-oh,fmoc-p-bromo-phe-oh,fmoc-d-4-bromophenylalanine,fmoc-l-4-bromophe,fmoc-l-phe 4-br-oh,fmoc-4'-bromo-l-phe
2734449
(2S)-3-(4-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Br)C(=O)O
198561-04-5
C24H20BrNO4
466.331
MFCD00273460
TVBAVBWXRDHONF-QFIPXVFZSA-N
fmoc-l-4-bromophenylalanine,s-n-fmoc-4-bromophenylalanine,fmoc-phe 4-br-oh,fmoc-4-bromo-l-phenylalanine,fmoc-l-4-br-phe-oh,fmoc-p-bromo-phe-oh,fmoc-d-4-bromophenylalanine,fmoc-l-4-bromophe,fmoc-l-phe 4-br-oh,fmoc-4'-bromo-l-phe
2734449
(2S)-3-(4-bromophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Br)C(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 466.32
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-4-Bromophenylalanine, 95%, (98% e.e.)
Show More Show Less