Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-N-FMOC-4-Chlorophenylalanine, 95%, 98% e.e., ACROS Organics™

Click to view available options
Quantity:
1g
175453-08-4
C24H20ClNO4
421.877
MFCD00080281
CQPNKLNINBUUOM-QFIPXVFZSA-N
fmoc-4-chloro-l-phenylalanine,s-n-fmoc-4-chlorophenylalanine,fmoc-phe 4-cl-oh,fmoc-l-4-chlorophenylalanine,fmoc-l-phe 4-cl-oh,fmoc-4-chloro-phe-oh,fmoc-l-4-chlorophe,2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid,phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
2734471
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
175453-08-4
C24H20ClNO4
421.877
MFCD00080281
CQPNKLNINBUUOM-QFIPXVFZSA-N
fmoc-4-chloro-l-phenylalanine,s-n-fmoc-4-chlorophenylalanine,fmoc-phe 4-cl-oh,fmoc-l-4-chlorophenylalanine,fmoc-l-phe 4-cl-oh,fmoc-4-chloro-phe-oh,fmoc-l-4-chlorophe,2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid,phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
2734471
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
Infrared Spectrum Authentic
Quantity 1g
Color White
Enantiomeric Excess 97.5% min. (HPLC)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 421.87
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material (S)-N-FMOC-4-Chlorophenylalanine, 95%, (98% e.e.)
Show More Show Less